Sf-900™ III SFM
Gibco™

Sf-900™ III SFM

Sf-900™ III SFMは、加水分解物が少なく、無タンパク質で、動物由来成分を含まない培地で、細胞増殖における一貫性と信頼性とを提供します。この培地は動物由来成分を含まないため、迷入因子をスクリーニングする必要が少なくなります。バキュロウイルス発現系および安定昆虫発現系のどちらの系を使用した場合でも詳細を見る
製品番号(カタログ番号)数量
1265800120 L
12658019500 mL
126580271000 mL
1265803510 L
製品番号(カタログ番号) 12658001
価格(JPY)
244,400
Each
数量:
20 L
Customize this product
Sf-900™ III SFMは、加水分解物が少なく、無タンパク質で、動物由来成分を含まない培地で、細胞増殖における一貫性と信頼性とを提供します。この培地は動物由来成分を含まないため、迷入因子をスクリーニングする必要が少なくなります。

バキュロウイルス発現系および安定昆虫発現系のどちらの系を使用した場合でも、懸濁培養におけるSf9およびSf21細胞の増殖および組換え遺伝子発現をサポートおよび最大化するよう最適化されています。
Sf9細胞の増殖を10-14 x 106細胞/mlの密度までサポートします。
適切に保存した場合、12ヶ月間安定であることを保証します。

保存:
遮光した場所に2から8°Cで保存します。 適切に保存した場合、安定性は12ヶ月間保証されます。
研究用途にのみご使用ください。診断目的には使用できません。
仕様
細胞株Sf21, Sf9
細胞タイプInsect Cell
濃度1 X
培養環境Non-CO2
エンドトキシンレベルLow
製品ラインGibco, Sf-900
製品タイプInsect Cell Serum Free Medium (SFM)
純度または品質グレードCell Culture Grade
数量20 L
出荷条件Room Temperature
分類Animal Origin-free
Culture TypeSuspension Cell Culture
形状Liquid
Serum LevelSerum-free
無菌性Sterile
添加剤ありGlutamine
添加剤なしNo Phenol Red, No Sodium Pyruvate, No Sodium Bicarbonate
Unit SizeEach
組成および保存条件
Storage conditions: 2–8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 18 months from date of manufacture

よくあるご質問(FAQ)

What antimicrobials can be used in insect culture and at what concentration?

Many antibiotics are suitable for use with insect cells. The following antibiotics are commonly used:
- Penicillin/Streptomycine: 50-100 U/mL; 50-100 µg/mL
- Amphotericin B (Fungizone antimycotic): 0.25 µg/mL
- Gentamicin: 0.5 mL of 10 mg/mL solution in 500 mL media (final concentration: 10 µg/mL)

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Is it necessary to heat-inactivate my serum before adding it to my medium?

Heat inactivation is not necessary. Our team has routinely used serum that has not been heat-inactivated, and we have not observed any effect on cell growth or morphology.

Many cells do not require heat-inactivated FBS. Some cells prefer heat-inactivated FBS. For instance, we use heat-inactivated FBS for our insect cell lines, i.e., Sf9 and Sf21 cells.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Do you offer serum-free insect media?

Yes, we offer a variety of serum-free insect media. Please go here (http://www.thermofisher.com/us/en/home/life-science/cell-culture/insect-cell-culture/insect-cell-culture-misc/serum-free-media.html?icid=cvc-insect-media-c2t2) to view the media we offer and the differences between them.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.