3-Methylcyclohexanone, 97%
3-Methylcyclohexanone, 97%
3-Methylcyclohexanone, 97%
Thermo Scientific Chemicals

3-Methylcyclohexanone, 97%

CAS: 591-24-2 | C7H12O | 112.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 126631000
Price (EUR)
59,65
Online exclusive
65,80
Save 6,15 (9%)
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
59,65
Online exclusive
65,80
Save 6,15 (9%)
Each
Chemical Identifiers
CAS591-24-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.4440 to 1.4460 (20°C, 589 nm)
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only