3-Methyl-2-cyclohexen-1-one, 98%
3-Methyl-2-cyclohexen-1-one, 98%
3-Methyl-2-cyclohexen-1-one, 98%
Thermo Scientific Chemicals

3-Methyl-2-cyclohexen-1-one, 98%

CAS: 1193-18-6 | C7H10O | 110.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 126660500
Price (USD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1193-18-6
IUPAC Name3-methylcyclohex-2-en-1-one
Molecular FormulaC7H10O
InChI KeyIITQJMYAYSNIMI-UHFFFAOYSA-N
SMILESCC1=CC(=O)CCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=97.5 %
Appearance (Color)Clear colorless to light yellow to brown
Infrared spectrumConforms
Refractive index1.4935 to 1.4955 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only