N-Methyldiethanolamine, 99+%, Thermo Scientific Chemicals
N-Methyldiethanolamine, 99+%, Thermo Scientific Chemicals
N-Methyldiethanolamine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Methyldiethanolamine, 99+%, Thermo Scientific Chemicals

数量:
250 g
1 kg
2.5 kg
パッケージ:
Glass bottle
製品番号(カタログ番号) 126722500
価格(JPY)
-
数量:
250 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS105-59-9
IUPAC Name2-[(2-hydroxyethyl)(methyl)amino]ethan-1-ol
Molecular FormulaC5H13NO2
InChI KeyCRVGTESFCCXCTH-UHFFFAOYSA-N
SMILESCN(CCO)CCO
さらに表示
Appearance (Form)Liquid
Refractive index1.4675 to 1.4695 (20°C, 589 nm)
Infrared spectrumConforms
Water=<0.5 %
Appearance (Color)Clear colorless to light yellow
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only