2-Methylfuran, 99%, stabilized
2-Methylfuran, 99%, stabilized
2-Methylfuran, 99%, stabilized
Thermo Scientific Chemicals

2-Methylfuran, 99%, stabilized

CAS: 534-22-5 | C5H6O | 82.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
250 mL
Packaging:
Glass bottle
Catalog number 126832500
Price (USD)
56.70
Each
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
56.70
Each
Chemical Identifiers
CAS534-22-5
IUPAC Name2-methylfuran
Molecular FormulaC5H6O
InChI KeyVQKFNUFAXTZWDK-UHFFFAOYSA-N
SMILESCC1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.3 %
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.4320 to 1.4340 (20°C, 589 nm)
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only