2-Methylhexane, 99%
2-Methylhexane, 99%
2-Methylhexane, 99%
Thermo Scientific Chemicals

2-Methylhexane, 99%

CAS: 591-76-4 | C7H16 | 100.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 126910050
Price (USD)
-
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS591-76-4
IUPAC Name2-methylhexane
Molecular FormulaC7H16
InChI KeyGXDHCNNESPLIKD-UHFFFAOYSA-N
SMILESCCCCC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless
GC>=98.5 %
Appearance (Form)Liquid
Refractive index1.3835 to 1.3855 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only