1-Methylimidazole, 99%
1-Methylimidazole, 99%
1-Methylimidazole, 99%
Thermo Scientific Chemicals

1-Methylimidazole, 99%

CAS: 616-47-7 | C4H6N2 | 82.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
5 kg
Packaging:
View all
Plastic drum
Catalog number 126990050
Price (USD)
726.65
Online Exclusive
807.00
Save 80.35 (10%)
Each
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
726.65
Online Exclusive
807.00
Save 80.35 (10%)
Each
Chemical Identifiers
CAS616-47-7
IUPAC Name1-methyl-1H-imidazole
Molecular FormulaC4H6N2
InChI KeyMCTWTZJPVLRJOU-UHFFFAOYSA-N
SMILESCN1C=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Water=<0.5 % (K.F.)
Appearance (Form)Liquid
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only