2-metilimidazol, 99 %, Thermo Scientific Chemicals
2-metilimidazol, 99 %, Thermo Scientific Chemicals
2-metilimidazol, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-metilimidazol, 99 %, Thermo Scientific Chemicals

CAS: 693-98-1 | C4H6N2 | 82.11 g/mol
Have Questions?
Número de catálogoCantidadEnvase
1270000101 kgFrasco de plástico
Número de catálogo 127000010
Precio (USD)
-
Cantidad:
1 kg
Envase:
Frasco de plástico
Pedido a granel o personalizado
Identificadores químicos
CAS693-98-1
IUPAC Name2-methyl-1H-imidazole
Molecular FormulaC4H6N2
InChI KeyLXBGSDVWAMZHDD-UHFFFAOYSA-N
SMILESCC1=NC=CN1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)White to light yellow
GC>=98.5 %
Melting point142°C to 145°C
Appearance (Form)Crystalline powder
Water=<0.5 % (K.F.)
Ver más
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only