1-Methylnaphthalene, 96%
1-Methylnaphthalene, 96%
1-Methylnaphthalene, 96%
Thermo Scientific Chemicals

1-Methylnaphthalene, 96%

CAS: 90-12-0 | C11H10 | 142.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 127161000
Price (EUR)
26,65
Online Exclusive
29,40
Save 2,75 (9%)
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
26,65
Online Exclusive
29,40
Save 2,75 (9%)
Each
Chemical Identifiers
CAS90-12-0
IUPAC Name1-methylnaphthalene
Molecular FormulaC11H10
InChI KeyQPUYECUOLPXSFR-UHFFFAOYSA-N
SMILESCC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=95 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow to light brown
Refractive index1.6100 to 1.6180 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only