2-Methylnaphthalene, 96%
2-Methylnaphthalene, 96%
2-Methylnaphthalene, 96%
Thermo Scientific Chemicals

2-Methylnaphthalene, 96%

CAS: 91-57-6 | C11H10 | 142.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 127175000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS91-57-6
IUPAC Name2-methylnaphthalene
Molecular FormulaC11H10
InChI KeyQIMMUPPBPVKWKM-UHFFFAOYSA-N
SMILESCC1=CC=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Low melting crystalline solid
Melting point32°C to 36°C
Appearance (Color)White
Infrared spectrumConforms
GC>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only