Methyl nicotinate, 99%
Methyl nicotinate, 99%
Methyl nicotinate, 99%
Thermo Scientific Chemicals

Methyl nicotinate, 99%

CAS: 93-60-7 | C7H7NO2 | 137.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 127215000
Price (TWD)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS93-60-7
IUPAC Namemethyl pyridine-3-carboxylate
Molecular FormulaC7H7NO2
InChI KeyYNBADRVTZLEFNH-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Melting point38°C to 43°C
Titration with HClO4>=98.5 %
Appearance (Color)White to brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only