Thermo Scientific Chemicals

2-Methylpentane, 99+%, pure, Thermo Scientific Chemicals

Catalog number: 127340010
1 L, Glass bottle, Each
Thermo Scientific Chemicals

2-Methylpentane, 99+%, pure, Thermo Scientific Chemicals

Catalog number: 127340010
1 L, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
107-83-5
IUPAC Name
2-methylpentane
Molecular Formula
C6H14
InChI Key
AFABGHUZZDYHJO-UHFFFAOYSA-N
SMILES
CCCC(C)C
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless
GC
>=99.0 %
Appearance (Form)
Liquid
Refractive index
1.3700 to 1.3720 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text