N-Methylpiperidine, 99%
N-Methylpiperidine, 99%
N-Methylpiperidine, 99%
Thermo Scientific Chemicals

N-Methylpiperidine, 99%

CAS: 626-67-5 | C6H14ClN | 135.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number 127480010
Price (USD)
226.65
Online Exclusive
252.00
Save 25.35 (10%)
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
226.65
Online Exclusive
252.00
Save 25.35 (10%)
Each
Chemical Identifiers
CAS626-67-5
IUPAC Namehydrogen 1-methylpiperidine chloride
Molecular FormulaC6H14ClN
InChI KeyQDUXDCXILAPLAG-UHFFFAOYSA-N
SMILES[H+].[Cl-].CN1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only