4-Methylpiperidine, 99%
4-Methylpiperidine, 99%
4-Methylpiperidine, 99%
Thermo Scientific Chemicals

4-Methylpiperidine, 99%

CAS: 626-58-4 | C6H13N | 99.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Catalog number 127515000
Price (USD)
136.65
Special offer
Online exclusive
Ends: 28-Dec-2026
160.00
Save 23.35
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
136.65
Special offer
Online exclusive
Ends: 28-Dec-2026
160.00
Save 23.35
Each
Chemical Identifiers
CAS626-58-4
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4440 to 1.4480 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only