p-Methylstyrene, 97%, stabilized
p-Methylstyrene, 97%, stabilized
p-Methylstyrene, 97%, stabilized
Thermo Scientific Chemicals

p-Methylstyrene, 97%, stabilized

CAS: 622-97-9 | C9H10 | 118.18 g/mol
数量:
10 mL
100 mL
500 mL
2.5 L
パッケージ:
Glass bottle
製品番号(カタログ番号) 127700025
価格(JPY)
-
数量:
2.5 L
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS622-97-9
IUPAC Name1-ethenyl-4-methylbenzene
Molecular FormulaC9H10
InChI KeyJLBJTVDPSNHSKJ-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C)C=C1
さらに表示
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=96.0 %
Refractive index1.5410 to 1.5430 (20°C, 589 nm)
Infrared spectrumConforms
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only