p-Methylstyrene, 97%, stabilized
p-Methylstyrene, 97%, stabilized
p-Methylstyrene, 97%, stabilized
Thermo Scientific Chemicals

p-Methylstyrene, 97%, stabilized

CAS: 622-97-9 | C9H10 | 118.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 127700100
Price (USD)
-
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS622-97-9
IUPAC Name1-ethenyl-4-methylbenzene
Molecular FormulaC9H10
InChI KeyJLBJTVDPSNHSKJ-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=96.0 %
Refractive index1.5410 to 1.5430 (20°C, 589 nm)
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only