Luria Broth Base (Miller's LB Broth Base), powder
Invitrogen™

Luria Broth Base (Miller's LB Broth Base), powder

Luria broth (LB) is a popular, rich medium used to cultivate bacteria and to perform coliphage plaque assays. The original자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
12795027500 g
127950842.5 kg
카탈로그 번호 12795027
제품 가격(KRW)
144,000
온라인 행사
Ends: 31-Dec-2026
164,000
할인액 20,000
Each
수량:
500 g
제품 가격(KRW)
144,000
온라인 행사
Ends: 31-Dec-2026
164,000
할인액 20,000
Each

Luria broth (LB) is a popular, rich medium used to cultivate bacteria and to perform coliphage plaque assays. The original formulation published in 1951 by Guiseppe Bertani to study phage plaque formation and named Lysogeny broth. It is also known as Lennox broth and Luria-Bertani broth.

Widely used LB medium formulations differ in salt concentration:
Miller: 10 g/L NaCl
Lennox: 5 g/L NaCl
Luria: 0.5 g/L NaCl

Luria Broth Base (Miller's formulation) is recommended for the general maintenance and growth of E. coli as it allows for fast cell expansion and supports high cell density. In cases where salt-sensitive antibiotics are being used for selection (such as Zeocin, tetracycline or chloramphenicol), Lennox formulation medium would be recommended.

Luria Broth Base formulation
Formulation per liter: 10 g SELECT Peptone 140, 5 g SELECT Yeast Extract, and 10 g NaCl.
To prepare 1 L of Miller-LB media, add 25 g of powder to 1 L distilled water and mix by vortexing. Adjust the pH to 7.0 with 5 N NaOH and sterilize by autoclaving for 25 minutes at 121°C.

Find the media format that fits your needs
We offer other ready-to-use media formulations, such as LB Broth, Terrific Broth or S.O.C recovery medium.
Explore other media formats for bacterial growth.

For Research Use Only. Not for use in diagnostic procedures.
사양
형식Bottle
미디어 유형LB Broth Base (Miller's LB Broth Base)
준비 방법Autoclave
수량500 g
배송 조건Room Temperature
제품 유형Dehydrated Culture Media
종Escherichia coli
타겟 유기체 분류Coliforms
Unit SizeEach
구성 및 보관
• 500 g LB Dehydrated Media Powder

Store at room temperature.

자주 묻는 질문(FAQ)

Do you offer an alternative for Luria Broth Base (Miller's LB Broth Base), powder (Cat. No. 12795027, 12795084)?

We do not offer any alternatives for this product.