Methylurea, 97%
Methylurea, 97%
Methylurea, 97%
Thermo Scientific Chemicals

Methylurea, 97%

CAS: 598-50-5 | C2H6N2O | 74.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 127995000
Price (TWD)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS598-50-5
IUPAC Namemethylurea
Molecular FormulaC2H6N2O
InChI KeyXGEGHDBEHXKFPX-UHFFFAOYSA-N
SMILESCNC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline solid
Titration with HClO4>=96.0 % (On dry substance)
Appearance (Color)White to off-white
Water=<2 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only