Mucochloric acid, 99%
Mucochloric acid, 99%
Mucochloric acid, 99%
Acros Organics

Mucochloric acid, 99%

CAS: 87-56-9 | C4H2Cl2O3 | 168.96 g/mol
Have Questions?
Catalog NumberQuantity
128061000
also known as 12806
100 g
Catalog number 128061000
also known as 12806
HSN 29183090
Price (INR)
5,728.00
Each
Quantity:
100 g
Request bulk or custom format
Price (INR)
5,728.00
Each
Chemical Identifiers
CAS87-56-9
IUPAC Name(2Z)-2,3-dichloro-4-oxobut-2-enoic acid
Molecular FormulaC4H2Cl2O3
InChI KeyLUMLZKVIXLWTCI-IHWYPQMZSA-N
SMILESOC(=O)C(\Cl)=C(\Cl)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point125°C to 128°C
Appearance (Color)White to light beige
Infrared spectrumConforms
Titration with NaOH>=98.5 % (on dry substance)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only