1-Naphthaldehyde, 95%
1-Naphthaldehyde, 95%
1-Naphthaldehyde, 95%
Thermo Scientific Chemicals

1-Naphthaldehyde, 95%

CAS: 66-77-3 | C11H8O | 156.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 128091000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS66-77-3
IUPAC Namenaphthalene-1-carbaldehyde
Molecular FormulaC11H8O
InChI KeySQAINHDHICKHLX-UHFFFAOYSA-N
SMILESO=CC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=94 %
Appearance (Color)Clear yellow to yellow-brown
Refractive index1.6500 to 1.6540 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only