1,8-Naphthalic anhydride, 97%
1,8-Naphthalic anhydride, 97%
1,8-Naphthalic anhydride, 97%
Thermo Scientific Chemicals

1,8-Naphthalic anhydride, 97%

CAS: 81-84-5 | C12H6O3 | 198.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Catalog number 128140025
Price (USD)
390.00
Each
Quantity:
2.5 kg
Request bulk or custom format
Price (USD)
390.00
Each
Chemical Identifiers
CAS81-84-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige to light brown
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Form)Powder
Melting point>=269°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only