Neopentyl alcohol, 99%
Neopentyl alcohol, 99%
Neopentyl alcohol, 99%
Thermo Scientific Chemicals

Neopentyl alcohol, 99%

CAS: 75-84-3 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 128252500
Price (EUR)
317,00
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
317,00
Each
Chemical Identifiers
CAS75-84-3
IUPAC Name2,2-dimethylpropan-1-ol
Molecular FormulaC5H12O
InChI KeyKPSSIOMAKSHJJG-UHFFFAOYSA-N
SMILESCC(C)(C)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Waxy crystalline solid
GC>=98.5 %
Appearance (Color)Colorless
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only