p-Nitroacetophenone, 97%
p-Nitroacetophenone, 97%
p-Nitroacetophenone, 97%
Thermo Scientific Chemicals

p-Nitroacetophenone, 97%

CAS: 100-19-6 | C8H7NO3 | 165.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
1 kg
Packaging:
Glass bottle
Catalog number 128341000
Price (USD)
49.65
Online exclusive
55.30
Save 5.65
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
49.65
Online exclusive
55.30
Save 5.65
Each
Chemical Identifiers
CAS100-19-6
IUPAC Name1-(4-nitrophenyl)ethan-1-one
Molecular FormulaC8H7NO3
InChI KeyYQYGPGKTNQNXMH-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Free acid=<0.2 % (as 4-Nitrobenzoic acid)
Appearance (Form)Crystalline powder
Melting point76°C to 80°C
Water=<0.6 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only