3-Nitroaniline, 98%
3-Nitroaniline, 98%
3-Nitroaniline, 98%
Thermo Scientific Chemicals

3-Nitroaniline, 98%

CAS: 99-09-2 | C6H6N2O2 | 138.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 128365000
Price (USD)
127.65
Online exclusive
142.00
Save 14.35
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
127.65
Online exclusive
142.00
Save 14.35
Each
Chemical Identifiers
CAS99-09-2
IUPAC Name3-nitroaniline
Molecular FormulaC6H6N2O2
InChI KeyXJCVRTZCHMZPBD-UHFFFAOYSA-N
SMILESNC1=CC=CC(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to ochre to orange
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Crystalline powder or crystals and/or chunks
Melting point111°C to 114°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only