4-Nitroanilina, 99 %, Thermo Scientific Chemicals
4-Nitroanilina, 99 %, Thermo Scientific Chemicals
4-Nitroanilina, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Nitroanilina, 99 %, Thermo Scientific Chemicals

CAS: 100-01-6 | C6H6N2O2 | 138.13 g/mol
Have Questions?
Número de catálogoCantidadEnvase
128371000100 gFrasco de vidrio
Número de catálogo 128371000
Precio (USD)
-
Cantidad:
100 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS100-01-6
EspecificacionesSpecification SheetHoja de especificaciones
GC>=98.5 %
Appearance (Form)Crystalline powder or crystals or flakes
Infrared spectrumConforms
Melting point147°C to 151°C
Appearance (Color)Yellow to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicador
RUO – Research Use Only