Nitrobenzene, 99%, Extra Pure
Nitrobenzene, 99%, Extra Pure
Nitrobenzene, 99%, Extra Pure
Thermo Scientific Chemicals

Nitrobenzene, 99%, Extra Pure

CAS: 98-95-3 | C6H5NO2 | 123.11 g/mol
数量:
2.5 L
10 L
パッケージ:
すべて表示
Glass bottle
製品番号(カタログ番号) 128420025
価格(JPY)
-
数量:
2.5 L
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS98-95-3
IUPAC Namenitrobenzene
Molecular FormulaC6H5NO2
InChI KeyLQNUZADURLCDLV-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC=C1
さらに表示
Infrared spectrumConforms
Water=<0.15 %
Appearance (Form)Liquid
Refractive index1.5500 to 1.5540 (20°C, 589 nm)
Appearance (Color)Clear yellow
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only