4-Nitrobenzoic acid, 99+%
4-Nitrobenzoic acid, 99+%
4-Nitrobenzoic acid, 99+%
Thermo Scientific Chemicals

4-Nitrobenzoic acid, 99+%

CAS: 62-23-7 | C7H5NO4 | 167.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
1 kg
Packaging:
Plastic bottle
Catalog number 128460250
Price (USD)
19.65
Special offer
Online exclusive
Ends: 28-Dec-2026
23.30
Save 3.65
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
19.65
Special offer
Online exclusive
Ends: 28-Dec-2026
23.30
Save 3.65
Each
Chemical Identifiers
CAS62-23-7
IUPAC Name4-nitrobenzoic acid
Molecular FormulaC7H5NO4
InChI KeyOTLNPYWUJOZPPA-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point238°C to 242°C
Appearance (Form)Powder or crystalline powder or crystals
Titration with NaOH99.0 to 101.0 %
Infrared spectrumConforms
HPLC>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only