4-Nitrobenzonitrile, 97%
4-Nitrobenzonitrile, 97%
4-Nitrobenzonitrile, 97%
Thermo Scientific Chemicals

4-Nitrobenzonitrile, 97%

CAS: 619-72-7 | C7H4N2O2 | 148.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 128481000
Price (USD)
185.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
185.00
Each
Chemical Identifiers
CAS619-72-7
IUPAC Name4-nitrobenzonitrile
Molecular FormulaC7H4N2O2
InChI KeyNKJIFDNZPGLLSH-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point144°C to 149°C
GC>=96.0 %
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Appearance (Color)Yellow to beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only