4-(4-Nitrobenzyl)pyridine, 98%
4-(4-Nitrobenzyl)pyridine, 98%
4-(4-Nitrobenzyl)pyridine, 98%
Thermo Scientific Chemicals

4-(4-Nitrobenzyl)pyridine, 98%

CAS: 1083-48-3 | C12H10N2O2 | 214.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 128550100
Price (EUR)
47,30
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
47,30
Each
Chemical Identifiers
CAS1083-48-3
IUPAC Name4-[(4-nitrophenyl)methyl]pyridine
Molecular FormulaC12H10N2O2
InChI KeyMNHKUCBXXMFQDM-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(CC2=CC=NC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Appearance (Color)Light yellow
Melting point69°C to 72°C
Titration with HClO4>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only