4-Nitrophenylacetic acid, 99%
4-Nitrophenylacetic acid, 99%
4-Nitrophenylacetic acid, 99%
Thermo Scientific Chemicals

4-Nitrophenylacetic acid, 99%

CAS: 104-03-0 | C8H7NO4 | 181.15 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
50 g
100 g
產品號碼 128761000
價格 (TWD)
-
Quantity:
100 g
Request bulk or custom format
化學識別
CAS104-03-0
IUPAC Name2-(4-nitrophenyl)acetic acid
Molecular FormulaC8H7NO4
InChI KeyYBADLXQNJCMBKR-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC=C(C=C1)[N+]([O-])=O
檢視更多
規格Specification Sheet規格表
Appearance (Color)Yellow to beige
Titration with NaOH>=98.5 %
Melting point153°C to 156°C
Infrared spectrumConforms
HPLC>=98.5 %
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only