Thermo Scientific™
I-Chem™ Boston Round Narrow-Mouth Clear Glass Bottles with Closure
Catalog number: 129-0500
Thermo Scientific™
I-Chem™ Boston Round Narrow-Mouth Clear Glass Bottles with Closure
Catalog number: 129-0500
Choose this Thermo Scientific™ I-Chem™ Boston Round Narrow-Mouth Clear Glass Bottle for collecting water samples for a variety of organic and inorganic target analytes where light protection is not required. Bottles are available Certified to meet EPA performance-based specifications for semi-volatile organics, pesticides, PCBs and metals anlyses, Processed the same way as the certified bottles but without certification documentation, or Unprocessed and ready for your own cleaning procedure.
Catalog Number
129-0500
Unit Size
Case of 12
Capacity (Metric)
500 mL
Level
Unprocessed
dimClosureSize
No. per Case
12
Price (USD)
Price: 80.60
Your price:
Availability
-
Full specifications
Quantity
Closure MaterialPolypropylene
Closure Size28-400 mm
Closure TypeClosed-top Cap
Diameter (Metric)74 mm
Height (English)6.61 in.
Height (Metric)168 mm
Product TypeJar
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
Certification ParametersNone
ColorClear
DescriptionClear Glass Boston Round
Diameter (English) Inner0.75 in.
Diameter (English)2.91 in.
Diameter (Metric) Inner1.9 cm
MaterialGlass (Type 3)
No. per Case12
Replacement ClosureB167-2800BL
Closure ColorWhite
LevelUnprocessed
Liner MaterialPTFE
Volume (English) Brim16.7 oz.
Volume (Metric) Brim493 mL
Certifications/ComplianceNone
MouthNarrow
Unit SizeCase of 12
Catalog Number
Unit Size
Case of 12
Capacity (Metric)
125 mL
Level
Unprocessed
dimClosureSize
No. per Case
12
Price (USD)
Availability
-
Full specifications
Quantity
-
Closure MaterialPolypropylene
Closure Size22-400 mm
Closure TypeClosed-top Cap
Diameter (Metric)48 mm
Height (English)4.4 in.
Height (Metric)112 mm
Product TypeJar
Capacity4 oz., 125 mL
Capacity (Metric)125 mL
Capacity (English)4 oz.
Certification ParametersNone
ColorClear
DescriptionClear Glass Boston Round
Diameter (English) Inner0.56 in.
Diameter (English)1.88 in.
Diameter (Metric) Inner1.4 cm
MaterialGlass (Type 3)
No. per Case12
Replacement ClosureB267-2200UB
Closure ColorWhite
LevelUnprocessed
Liner MaterialPTFE
Volume (English) Brim4.3 oz.
Volume (Metric) Brim128 mL
Certifications/ComplianceNone
MouthNarrow
Unit SizeCase of 12
Catalog Number
Unit Size
Case of 12
Capacity (Metric)
125 mL
Level
Certified
dimClosureSize
No. per Case
12
Price (USD)
Price: 63.50
Your price:
Availability
-
Full specifications
Quantity
Closure MaterialPolypropylene
Closure Size22-400 mm
Closure TypeClosed-top Cap
Diameter (Metric)48 mm
Height (English)4.4 in.
Height (Metric)112 mm
Product TypeJar
Capacity4 oz., 125 mL
Capacity (Metric)125 mL
Capacity (English)4 oz.
Certification ParametersSemi-VOA, pesticides, PCBs
ColorClear
DescriptionClear Glass Boston Round
Diameter (English) Inner0.56 in.
Diameter (English)1.88 in.
Diameter (Metric) Inner1.4 cm
MaterialGlass (Type 3)
No. per Case12
Replacement ClosureB267-2200UB
Closure ColorWhite
LevelCertified
Liner MaterialPTFE
Volume (English) Brim4.3 oz.
Volume (Metric) Brim128 mL
Certifications/ComplianceCertified and Processed bottles meet or exceed EPA performance based standards for semi-volatile organics, pesticides, and PCB's analysis; manufactured under an ISO certified quality management system. Certificate of Analysis (CofA) included.
MouthNarrow
Unit SizeCase of 12
Catalog Number
Unit Size
Case of 12
Capacity (Metric)
125 mL
Level
Processed
dimClosureSize
No. per Case
12
Price (USD)
Price: 54.90
Your price:
Availability
-
Full specifications
Quantity
Closure MaterialPolypropylene
Closure Size22-400 mm
Closure TypeClosed-top Cap
Diameter (Metric)48 mm
Height (English)4.4 in.
Height (Metric)112 mm
Product TypeJar
Capacity4 oz., 125 mL
Capacity (Metric)125 mL
Capacity (English)4 oz.
Certification ParametersSemi-VOA, pesticides, PCBs
ColorClear
DescriptionClear Glass Boston Round
Diameter (English) Inner0.56 in.
Diameter (English)1.88 in.
Diameter (Metric) Inner1.4 cm
MaterialGlass (Type 3)
No. per Case12
Replacement ClosureB267-2200UB
Closure ColorWhite
LevelProcessed
Liner MaterialPTFE
Volume (English) Brim4.3 oz.
Volume (Metric) Brim128 mL
Certifications/ComplianceCertified and Processed bottles meet or exceed EPA performance based standards for semi-volatile organics, pesticides, and PCB's analysis; manufactured under an ISO certified quality management system.
MouthNarrow
Unit SizeCase of 12
Catalog Number
Unit Size
Case of 12
Capacity (Metric)
250 mL
Level
Unprocessed
dimClosureSize
No. per Case
12
Price (USD)
Price: 46.20
Your price:
Availability
-
Full specifications
Quantity
Closure MaterialPolypropylene
Closure Size24-414 mm
Closure TypeClosed-top Cap
Diameter (Metric)60 mm
Height (English)5.35 in.
Height (Metric)136 mm
Product TypeJar
Capacity8 oz., 250 mL
Capacity (Metric)250 mL
Capacity (English)8 oz.
Certification ParametersNone
ColorClear
DescriptionClear Glass Boston Round
Diameter (English) Inner0.63 in.
Diameter (English)2.36 in.
Diameter (Metric) Inner1.6 cm
MaterialGlass (Type 3)
No. per Case12
Replacement ClosureB267-2414UBB
Closure ColorWhite
LevelUnprocessed
Liner MaterialPTFE
Volume (English) Brim8.5 oz.
Volume (Metric) Brim250 mL
Certifications/ComplianceNone
MouthNarrow
Unit SizeCase of 12
Catalog Number
Unit Size
Case of 12
Capacity (Metric)
250 mL
Level
Certified
dimClosureSize
No. per Case
12
Price (USD)
Price: 60.80
Your price:
Availability
-
Full specifications
Quantity
Closure MaterialPolypropylene
Closure Size24-414 mm
Closure TypeClosed-top Cap
Diameter (Metric)60 mm
Height (English)5.35 in.
Height (Metric)136 mm
Product TypeJar
Capacity8 oz., 250 mL
Capacity (Metric)250 mL
Capacity (English)8 oz.
Certification ParametersSemi-VOA, pesticides, PCBs
ColorClear
DescriptionClear Glass Boston Round
Diameter (English) Inner0.63 in.
Diameter (English)2.36 in.
Diameter (Metric) Inner1.6 cm
MaterialGlass (Type 3)
No. per Case12
Replacement ClosureB267-2414UBB
Closure ColorWhite
LevelCertified
Liner MaterialPTFE
Volume (English) Brim8.5 oz.
Volume (Metric) Brim250 mL
Certifications/ComplianceCertified and Processed bottles meet or exceed EPA performance based standards for semi-volatile organics, pesticides, and PCB's analysis; manufactured under an ISO certified quality management system. Certificate of Analysis (CofA) included.
MouthNarrow
Unit SizeCase of 12
Catalog Number
Unit Size
Case of 12
Capacity (Metric)
250 mL
Level
Processed
dimClosureSize
No. per Case
12
Price (USD)
Price: 52.30
Your price:
Availability
-
Full specifications
Quantity
Closure MaterialPolypropylene
Closure Size24-414 mm
Closure TypeClosed-top Cap
Diameter (Metric)60 mm
Height (English)5.35 in.
Height (Metric)136 mm
Product TypeJar
Capacity8 oz., 250 mL
Capacity (Metric)250 mL
Capacity (English)8 oz.
Certification ParametersSemi-VOA, pesticides, PCBs
ColorClear
DescriptionClear Glass Boston Round
Diameter (English) Inner0.63 in.
Diameter (English)2.36 in.
Diameter (Metric) Inner1.6 cm
MaterialGlass (Type 3)
No. per Case12
Replacement ClosureB267-2414UBB
Closure ColorWhite
LevelProcessed
Liner MaterialPTFE
Volume (English) Brim8.5 oz.
Volume (Metric) Brim250 mL
Certifications/ComplianceCertified and Processed bottles meet or exceed EPA performance based standards for semi-volatile organics, pesticides, and PCB's analysis; manufactured under an ISO certified quality management system.
MouthNarrow
Unit SizeCase of 12
Catalog Number
Unit Size
Case of 12
Capacity (Metric)
500 mL
Level
Processed
dimClosureSize
No. per Case
12
Price (USD)
Price: 80.60
Your price:
Availability
-
Full specifications
Quantity
Closure MaterialPolypropylene
Closure Size28-400 mm
Closure TypeClosed-top Cap
Diameter (Metric)74 mm
Height (English)6.61 in.
Height (Metric)168 mm
Product TypeJar
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
Certification ParametersSemi-VOA, pesticides, PCBs
ColorClear
DescriptionClear Glass Boston Round
Diameter (English) Inner0.75 in.
Diameter (English)2.91 in.
Diameter (Metric) Inner1.9 cm
MaterialGlass (Type 3)
No. per Case12
Replacement ClosureB167-2800BL
Closure ColorWhite
LevelProcessed
Liner MaterialPTFE
Volume (English) Brim16.7 oz.
Volume (Metric) Brim493 mL
Certifications/ComplianceCertified and Processed bottles meet or exceed EPA performance based standards for semi-volatile organics, pesticides, and PCB's analysis; manufactured under an ISO certified quality management system.
MouthNarrow
Unit SizeCase of 12
Catalog Number
Unit Size
Case of 12
Capacity (Metric)
500 mL
Level
Certified
dimClosureSize
No. per Case
12
Price (USD)
Price: 80.60
Your price:
Availability
-
Full specifications
Quantity
Closure MaterialPolypropylene
Closure Size28-400 mm
Closure TypeClosed-top Cap
Diameter (Metric)74 mm
Height (English)6.61 in.
Height (Metric)168 mm
Product TypeJar
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
Certification ParametersSemi-VOA, pesticides, PCBs
ColorClear
DescriptionClear Glass Boston Round
Diameter (English) Inner0.75 in.
Diameter (English)2.91 in.
Diameter (Metric) Inner1.9 cm
MaterialGlass (Type 3)
No. per Case12
Replacement ClosureB167-2800BL
Closure ColorWhite
LevelCertified
Liner MaterialPTFE
Volume (English) Brim16.7 oz.
Volume (Metric) Brim493 mL
Certifications/ComplianceCertified and Processed bottles meet or exceed EPA performance based standards for semi-volatile organics, pesticides, and PCB's analysis; manufactured under an ISO certified quality management system. Certificate of Analysis (CofA) included.
MouthNarrow
Unit SizeCase of 12
Catalog Number
Unit Size
Case of 12
Capacity (Metric)
950 mL
Level
Certified
dimClosureSize
No. per Case
12
Price (USD)
Price: 97.90
Your price:
Availability
-
Full specifications
Quantity
Closure MaterialPolypropylene
Closure Size33-400 mm
Closure TypeClosed-top Cap
Diameter (Metric)93 mm
Height (English)11.25 in.
Height (Metric)286 mm
Product TypeJar
Capacity32 oz., 950 mL
Capacity (Metric)950 mL
Capacity (English)32 oz.
Certification ParametersSemi-VOA, pesticides, PCBs, metals
ColorClear
DescriptionClear Glass Boston Round
Diameter (English) Inner0.88 in.
Diameter (English)3.66 in.
Diameter (Metric) Inner2.2 cm
MaterialGlass (Type 3)
No. per Case12
Replacement ClosureB167-3300BL
Closure ColorWhite
LevelCertified
Liner MaterialPTFE
Volume (English) Brim33 oz.
Volume (Metric) Brim975 mL
Certifications/ComplianceCertified and Processed bottles meet or exceed EPA performance based standards for semi-volatile organics, pesticides, PCB's and metals analysis; manufactured under an ISO certified quality management system. Certificate of Analysis (CofA) included.
MouthNarrow
Unit SizeCase of 12
Showing 10 of 12Show more
Catalog NumberSpecificationsUnit SizeCapacity (Metric)LeveldimClosureSizeNo. per CasePrice (USD)AvailabilityQuantity
129-0500Full specifications
Case of 12500 mLUnprocessed12
Price: 80.60
Your price:
-
Closure MaterialPolypropylene
Closure Size28-400 mm
Closure TypeClosed-top Cap
Diameter (Metric)74 mm
Height (English)6.61 in.
Height (Metric)168 mm
Product TypeJar
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
Certification ParametersNone
ColorClear
DescriptionClear Glass Boston Round
Diameter (English) Inner0.75 in.
Diameter (English)2.91 in.
Diameter (Metric) Inner1.9 cm
MaterialGlass (Type 3)
No. per Case12
Replacement ClosureB167-2800BL
Closure ColorWhite
LevelUnprocessed
Liner MaterialPTFE
Volume (English) Brim16.7 oz.
Volume (Metric) Brim493 mL
Certifications/ComplianceNone
MouthNarrow
Unit SizeCase of 12
129-0125Full specifications
Case of 12125 mLUnprocessed12Request A Quote--
Closure MaterialPolypropylene
Closure Size22-400 mm
Closure TypeClosed-top Cap
Diameter (Metric)48 mm
Height (English)4.4 in.
Height (Metric)112 mm
Product TypeJar
Capacity4 oz., 125 mL
Capacity (Metric)125 mL
Capacity (English)4 oz.
Certification ParametersNone
ColorClear
DescriptionClear Glass Boston Round
Diameter (English) Inner0.56 in.
Diameter (English)1.88 in.
Diameter (Metric) Inner1.4 cm
MaterialGlass (Type 3)
No. per Case12
Replacement ClosureB267-2200UB
Closure ColorWhite
LevelUnprocessed
Liner MaterialPTFE
Volume (English) Brim4.3 oz.
Volume (Metric) Brim128 mL
Certifications/ComplianceNone
MouthNarrow
Unit SizeCase of 12
329-0125Full specifications
Case of 12125 mLCertified12
Price: 63.50
Your price:
-
Closure MaterialPolypropylene
Closure Size22-400 mm
Closure TypeClosed-top Cap
Diameter (Metric)48 mm
Height (English)4.4 in.
Height (Metric)112 mm
Product TypeJar
Capacity4 oz., 125 mL
Capacity (Metric)125 mL
Capacity (English)4 oz.
Certification ParametersSemi-VOA, pesticides, PCBs
ColorClear
DescriptionClear Glass Boston Round
Diameter (English) Inner0.56 in.
Diameter (English)1.88 in.
Diameter (Metric) Inner1.4 cm
MaterialGlass (Type 3)
No. per Case12
Replacement ClosureB267-2200UB
Closure ColorWhite
LevelCertified
Liner MaterialPTFE
Volume (English) Brim4.3 oz.
Volume (Metric) Brim128 mL
Certifications/ComplianceCertified and Processed bottles meet or exceed EPA performance based standards for semi-volatile organics, pesticides, and PCB's analysis; manufactured under an ISO certified quality management system. Certificate of Analysis (CofA) included.
MouthNarrow
Unit SizeCase of 12
229-0125Full specifications
Case of 12125 mLProcessed12
Price: 54.90
Your price:
-
Closure MaterialPolypropylene
Closure Size22-400 mm
Closure TypeClosed-top Cap
Diameter (Metric)48 mm
Height (English)4.4 in.
Height (Metric)112 mm
Product TypeJar
Capacity4 oz., 125 mL
Capacity (Metric)125 mL
Capacity (English)4 oz.
Certification ParametersSemi-VOA, pesticides, PCBs
ColorClear
DescriptionClear Glass Boston Round
Diameter (English) Inner0.56 in.
Diameter (English)1.88 in.
Diameter (Metric) Inner1.4 cm
MaterialGlass (Type 3)
No. per Case12
Replacement ClosureB267-2200UB
Closure ColorWhite
LevelProcessed
Liner MaterialPTFE
Volume (English) Brim4.3 oz.
Volume (Metric) Brim128 mL
Certifications/ComplianceCertified and Processed bottles meet or exceed EPA performance based standards for semi-volatile organics, pesticides, and PCB's analysis; manufactured under an ISO certified quality management system.
MouthNarrow
Unit SizeCase of 12
129-0250Full specifications
Case of 12250 mLUnprocessed12
Price: 46.20
Your price:
-
Closure MaterialPolypropylene
Closure Size24-414 mm
Closure TypeClosed-top Cap
Diameter (Metric)60 mm
Height (English)5.35 in.
Height (Metric)136 mm
Product TypeJar
Capacity8 oz., 250 mL
Capacity (Metric)250 mL
Capacity (English)8 oz.
Certification ParametersNone
ColorClear
DescriptionClear Glass Boston Round
Diameter (English) Inner0.63 in.
Diameter (English)2.36 in.
Diameter (Metric) Inner1.6 cm
MaterialGlass (Type 3)
No. per Case12
Replacement ClosureB267-2414UBB
Closure ColorWhite
LevelUnprocessed
Liner MaterialPTFE
Volume (English) Brim8.5 oz.
Volume (Metric) Brim250 mL
Certifications/ComplianceNone
MouthNarrow
Unit SizeCase of 12
329-0250Full specifications
Case of 12250 mLCertified12
Price: 60.80
Your price:
-
Closure MaterialPolypropylene
Closure Size24-414 mm
Closure TypeClosed-top Cap
Diameter (Metric)60 mm
Height (English)5.35 in.
Height (Metric)136 mm
Product TypeJar
Capacity8 oz., 250 mL
Capacity (Metric)250 mL
Capacity (English)8 oz.
Certification ParametersSemi-VOA, pesticides, PCBs
ColorClear
DescriptionClear Glass Boston Round
Diameter (English) Inner0.63 in.
Diameter (English)2.36 in.
Diameter (Metric) Inner1.6 cm
MaterialGlass (Type 3)
No. per Case12
Replacement ClosureB267-2414UBB
Closure ColorWhite
LevelCertified
Liner MaterialPTFE
Volume (English) Brim8.5 oz.
Volume (Metric) Brim250 mL
Certifications/ComplianceCertified and Processed bottles meet or exceed EPA performance based standards for semi-volatile organics, pesticides, and PCB's analysis; manufactured under an ISO certified quality management system. Certificate of Analysis (CofA) included.
MouthNarrow
Unit SizeCase of 12
229-0250Full specifications
Case of 12250 mLProcessed12
Price: 52.30
Your price:
-
Closure MaterialPolypropylene
Closure Size24-414 mm
Closure TypeClosed-top Cap
Diameter (Metric)60 mm
Height (English)5.35 in.
Height (Metric)136 mm
Product TypeJar
Capacity8 oz., 250 mL
Capacity (Metric)250 mL
Capacity (English)8 oz.
Certification ParametersSemi-VOA, pesticides, PCBs
ColorClear
DescriptionClear Glass Boston Round
Diameter (English) Inner0.63 in.
Diameter (English)2.36 in.
Diameter (Metric) Inner1.6 cm
MaterialGlass (Type 3)
No. per Case12
Replacement ClosureB267-2414UBB
Closure ColorWhite
LevelProcessed
Liner MaterialPTFE
Volume (English) Brim8.5 oz.
Volume (Metric) Brim250 mL
Certifications/ComplianceCertified and Processed bottles meet or exceed EPA performance based standards for semi-volatile organics, pesticides, and PCB's analysis; manufactured under an ISO certified quality management system.
MouthNarrow
Unit SizeCase of 12
229-0500Full specifications
Case of 12500 mLProcessed12
Price: 80.60
Your price:
-
Closure MaterialPolypropylene
Closure Size28-400 mm
Closure TypeClosed-top Cap
Diameter (Metric)74 mm
Height (English)6.61 in.
Height (Metric)168 mm
Product TypeJar
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
Certification ParametersSemi-VOA, pesticides, PCBs
ColorClear
DescriptionClear Glass Boston Round
Diameter (English) Inner0.75 in.
Diameter (English)2.91 in.
Diameter (Metric) Inner1.9 cm
MaterialGlass (Type 3)
No. per Case12
Replacement ClosureB167-2800BL
Closure ColorWhite
LevelProcessed
Liner MaterialPTFE
Volume (English) Brim16.7 oz.
Volume (Metric) Brim493 mL
Certifications/ComplianceCertified and Processed bottles meet or exceed EPA performance based standards for semi-volatile organics, pesticides, and PCB's analysis; manufactured under an ISO certified quality management system.
MouthNarrow
Unit SizeCase of 12
329-0500Full specifications
Case of 12500 mLCertified12
Price: 80.60
Your price:
-
Closure MaterialPolypropylene
Closure Size28-400 mm
Closure TypeClosed-top Cap
Diameter (Metric)74 mm
Height (English)6.61 in.
Height (Metric)168 mm
Product TypeJar
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
Certification ParametersSemi-VOA, pesticides, PCBs
ColorClear
DescriptionClear Glass Boston Round
Diameter (English) Inner0.75 in.
Diameter (English)2.91 in.
Diameter (Metric) Inner1.9 cm
MaterialGlass (Type 3)
No. per Case12
Replacement ClosureB167-2800BL
Closure ColorWhite
LevelCertified
Liner MaterialPTFE
Volume (English) Brim16.7 oz.
Volume (Metric) Brim493 mL
Certifications/ComplianceCertified and Processed bottles meet or exceed EPA performance based standards for semi-volatile organics, pesticides, and PCB's analysis; manufactured under an ISO certified quality management system. Certificate of Analysis (CofA) included.
MouthNarrow
Unit SizeCase of 12
329-1000Full specifications
Case of 12950 mLCertified12
Price: 97.90
Your price:
-
Closure MaterialPolypropylene
Closure Size33-400 mm
Closure TypeClosed-top Cap
Diameter (Metric)93 mm
Height (English)11.25 in.
Height (Metric)286 mm
Product TypeJar
Capacity32 oz., 950 mL
Capacity (Metric)950 mL
Capacity (English)32 oz.
Certification ParametersSemi-VOA, pesticides, PCBs, metals
ColorClear
DescriptionClear Glass Boston Round
Diameter (English) Inner0.88 in.
Diameter (English)3.66 in.
Diameter (Metric) Inner2.2 cm
MaterialGlass (Type 3)
No. per Case12
Replacement ClosureB167-3300BL
Closure ColorWhite
LevelCertified
Liner MaterialPTFE
Volume (English) Brim33 oz.
Volume (Metric) Brim975 mL
Certifications/ComplianceCertified and Processed bottles meet or exceed EPA performance based standards for semi-volatile organics, pesticides, PCB's and metals analysis; manufactured under an ISO certified quality management system. Certificate of Analysis (CofA) included.
MouthNarrow
Unit SizeCase of 12
Showing 10 of 12Show more
  • Clear Type III soda-lime glass bottle
  • PTFE-lined white polypropylene closed-top cap
  • Sizes range from 125mL to 1000mL
  • Closures assembled on bottles
Certified bottles come with the following features:
  • Certified to meet or exceed EPA performance-based standards for semi-volatile organics, pesticides, PCBs and metals analyses
  • Certificate of Analysis (CofA) included in the case and barcoded with lot number for traceability
  • Each bottle individually barcoded with lot number and unique container number
  • Analysis labels included unattached for optional use
  • Carton sealed with security tape providing tamper evidence
  • Chipboard divider packaging to cushion and help prevent breakage; overpack required for shipping
Processed bottles are processed in the same manner as certified bottles and come with all the features of certified bottles, but bottles are not barcoded and CofA is not included.
Unprocessed bottles are ready for your own in-house cleaning and certification process. Boxed with chipboard dividers to help prevent breakage; overpack required for shipping.