Nitroterephthalic acid, 99+%
Nitroterephthalic acid, 99+%
Nitroterephthalic acid, 99+%
Thermo Scientific Chemicals

Nitroterephthalic acid, 99+%

CAS: 610-29-7 | C8H5NO6 | 211.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 129000250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS610-29-7
IUPAC Name2-nitrobenzene-1,4-dicarboxylic acid
Molecular FormulaC8H5NO6
InChI KeyQUMITRDILMWWBC-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(C(O)=O)C(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to light beige
Appearance (Form)Powder
Titration with NaOH>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only