n-Nonane, 99%, pure
n-Nonane, 99%, pure
n-Nonane, 99%, pure
Thermo Scientific Chemicals

n-Nonane, 99%, pure

CAS: 111-84-2 | C9H20 | 128.26 g/mol
数量:
100 mL
2.5 L
パッケージ:
Glass bottle
製品番号(カタログ番号) 129110025
価格(JPY)
-
見積もりを依頼する
数量:
2.5 L
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS111-84-2
IUPAC Namenonane
Molecular FormulaC9H20
InChI KeyBKIMMITUMNQMOS-UHFFFAOYSA-N
SMILESCCCCCCCCC
さらに表示
Appearance (Color)Clear colorless
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4040 to 1.4060 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only