1,9-Nonanediol, 98%
1,9-Nonanediol, 98%
1,9-Nonanediol, 98%
Thermo Scientific Chemicals

1,9-Nonanediol, 98%

CAS: 3937-56-2 | C9H20O2 | 160.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 129121000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3937-56-2
IUPAC Namenonane-1,9-diol
Molecular FormulaC9H20O2
InChI KeyALVZNPYWJMLXKV-UHFFFAOYSA-N
SMILESOCCCCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Melting point44°C to 48°C
Appearance (Form)Fused crystalline solid
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only