2-Octanone, 99+%
2-Octanone, 99+%
2-Octanone, 99+%
Thermo Scientific Chemicals

2-Octanone, 99+%

CAS: 111-13-7 | C8H16O | 128.21 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 mL
500 mL
2.5 L
Packaging:
Glass bottle
產品號碼 129420250
價格 (TWD)
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
規格
Assay Percent Range99% min. (GC)
Beilstein01, 704
CAS Max %100.0
CAS Min %99.0
CAS111-13-7
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only