1-Octen-3-ol, 98%
1-Octen-3-ol, 98%
1-Octen-3-ol, 98%
Thermo Scientific Chemicals

1-Octen-3-ol, 98%

CAS: 3391-86-4 | C8H16O | 128.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 129471000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3391-86-4
IUPAC Nameoct-1-en-3-ol
Molecular FormulaC8H16O
InChI KeyVSMOENVRRABVKN-UHFFFAOYNA-N
SMILESCCCCCC(O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Density(20°C) 0.832 to 0.842 kg/l
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only