Octyl aldehyde, 99%
Octyl aldehyde, 99%
Octyl aldehyde, 99%
Thermo Scientific Chemicals

Octyl aldehyde, 99%

CAS: 124-13-0 | C8H16O | 128.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 129481000
Price (USD)
41.65
Online exclusive
46.50
Save 4.85
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
41.65
Online exclusive
46.50
Save 4.85
Each
Chemical Identifiers
CAS124-13-0
IUPAC Nameoctanal
Molecular FormulaC8H16O
InChI KeyNUJGJRNETVAIRJ-UHFFFAOYSA-N
SMILESCCCCCCCC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4170 to 1.4200 (20°C, 589 nm)
GC>=98.5 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only