n-Octylamine, 99+%
n-Octylamine, 99+%
n-Octylamine, 99+%
Thermo Scientific Chemicals

n-Octylamine, 99+%

CAS: 111-86-4 | C8H19N | 129.24 g/mol
数量:
5 g
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 129490050
価格(JPY)
-
見積もりを依頼する
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS111-86-4
IUPAC Nameoctan-1-amine
Molecular FormulaC8H19N
InChI KeyIOQPZZOEVPZRBK-UHFFFAOYSA-N
SMILESCCCCCCCCN
さらに表示
GC>=99.0 %
Refractive index1.4280 to 1.4300 (20°C, 589 nm)
Appearance (Form)Clear liquid
Color scale=<20 APHA
Infrared spectrumConforms
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only