Thermo Scientific Chemicals

n-Octylamine, 99+%, Thermo Scientific Chemicals

Catalog number: 129495000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

n-Octylamine, 99+%, Thermo Scientific Chemicals

Catalog number: 129495000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 129495000
Price (USD)
Price: 93.20
Online price: 79.65
Your price:
Quantity
-

Chemical Identifiers

CAS
111-86-4
IUPAC Name
octan-1-amine
Molecular Formula
C8H19N
InChI Key
IOQPZZOEVPZRBK-UHFFFAOYSA-N
SMILES
CCCCCCCCN
Appearance (Form)
Clear liquid
GC
>=99.0 %
Refractive index
1.4280 to 1.4300 (20°C, 589 nm)
Color scale
=<20 APHA
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text