Oxamic acid, 98%
Oxamic acid, 98%
Oxamic acid, 98%
Thermo Scientific Chemicals

Oxamic acid, 98%

CAS: 471-47-6 | C2H3NO3 | 89.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 129621000
Price (EUR)
221,00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
221,00
Each
Chemical Identifiers
CAS471-47-6
IUPAC Namecarbamoylformic acid
Molecular FormulaC2H3NO3
InChI KeySOWBFZRMHSNYGE-UHFFFAOYSA-N
SMILESNC(=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only