Oxamide, 98%
Oxamide, 98%
Oxamide, 98%
Thermo Scientific Chemicals

Oxamide, 98%

CAS: 471-46-5 | C2H4N2O2 | 88.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 129655000
Price (USD)
130.65
Special offer
Online exclusive
Ends: 30-Jun-2026
154.00
Save 23.35 (15%)
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
130.65
Special offer
Online exclusive
Ends: 30-Jun-2026
154.00
Save 23.35 (15%)
Each
Chemical Identifiers
CAS471-46-5
IUPAC Nameethanediamide
Molecular FormulaC2H4N2O2
InChI KeyYIKSCQDJHCMVMK-UHFFFAOYSA-N
SMILESNC(=O)C(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Titration excess NaOH>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only