2-Oxazolidone, 98%
2-Oxazolidone, 98%
2-Oxazolidone, 98%
Thermo Scientific Chemicals

2-Oxazolidone, 98%

CAS: 497-25-6 | C3H5NO2 | 87.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 129660050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS497-25-6
IUPAC Name1,3-oxazolidin-2-one
Molecular FormulaC3H5NO2
InChI KeyIZXIZTKNFFYFOF-UHFFFAOYSA-N
SMILESO=C1NCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
GC>=97.5 %
Appearance (Color)Off-white to yellow to light beige
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only