Pamoic acid, 98+%
Pamoic acid, 98+%
Pamoic acid, 98+%
Thermo Scientific Chemicals

Pamoic acid, 98+%

CAS: 130-85-8 | C23H16O6 | 388.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 129721000
Price (USD)
99.65
Special offer
Online exclusive
Ends: 28-Dec-2026
117.00
Save 17.35
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
99.65
Special offer
Online exclusive
Ends: 28-Dec-2026
117.00
Save 17.35
Each
Chemical Identifiers
CAS130-85-8
IUPAC Name4-[(3-carboxy-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylic acid
Molecular FormulaC23H16O6
InChI KeyWLJNZVDCPSBLRP-UHFFFAOYSA-N
SMILESOC(=O)C1=C(O)C(CC2=C3C=CC=CC3=CC(C(O)=O)=C2O)=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to yellow-green
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Titration excess NaOH98.0 to 102.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only