1,4-Pentadiene, 99%
1,4-Pentadiene, 99%
1,4-Pentadiene, 99%
Acros Organics

1,4-Pentadiene, 99%

CAS: 591-93-5 | C5H8 | 68.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 129860050
HSN 29012990
Price (INR)
22,097.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
22,097.00
Each
Chemical Identifiers
CAS591-93-5
IUPAC Namepenta-1,4-diene
Molecular FormulaC5H8
InChI KeyQYZLKGVUSQXAMU-UHFFFAOYSA-N
SMILESC=CCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)Clear colorless
Refractive index1.3884 to 1.3904 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only