1,5-Pentanediol, 98%
1,5-Pentanediol, 98%
1,5-Pentanediol, 98%
Acros Organics

1,5-Pentanediol, 98%

CAS: 111-29-5 | C5H12O2 | 104.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
129955000
also known as 12995
500 gGlass bottle
Catalog number 129955000
also known as 12995
HSN 29053990
Price (INR)
7,859.00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
7,859.00
Each
Chemical Identifiers
CAS111-29-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Viscous liquid
Refractive index1.4490 to 1.4510 (20°C, 589 nm)
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only