2,4-Pentanedione, 99+%
2,4-Pentanedione, 99+%
2,4-Pentanedione, 99+%
Thermo Scientific Chemicals

2,4-Pentanedione, 99+%

CAS: 123-54-6 | C5H8O2 | 100.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 kg
2.5 kg
10 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 129960025
Price (USD)
-
Quantity:
2.5 kg
Request bulk or custom format
Chemical Identifiers
CAS123-54-6
IUPAC Namepentane-2,4-dione
Molecular FormulaC5H8O2
InChI KeyYRKCREAYFQTBPV-UHFFFAOYSA-N
SMILESCC(=O)CC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.1 %
Specific gravity(20°C) 0.972 to 0.978
Appearance (Form)Clear liquid
GC>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only