Thermo Scientific™
Samco™ 13 × 100mm Disposable Culture Tubes (DCTs)
Samco™ 13 × 100mm Disposable Culture Tubes (DCTs)
 Related applications:
Cell Culture Plastics
Choose the material that best suits your needs with Thermo Scientific™ Samco™ 13 × 100mm Disposable Culture Tubes, available in clarified polypropylene for strength and resistance, or polystyrene for superior clarity. Designed for most laboratory applications, these DCTs are chemical free and biologically inert. Each tube is precision molded and manufactured from premium grade materials. Our processes are monitored with quality checks which provide continued lot-to-lot uniformity and reliable performance. All sterile tubes are gamma-irradiated and ready to use.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Available with screw cap or without caps
  • Conveniently packaged—bulk or individually wrapped (sterile), and bulk or packs (non-sterile)
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Documents
Manuals & protocols