3,4,9,10-Perylenetetracarboxylic dianhydride, 98%
3,4,9,10-Perylenetetracarboxylic dianhydride, 98%
3,4,9,10-Perylenetetracarboxylic dianhydride, 98%
Thermo Scientific Chemicals

3,4,9,10-Perylenetetracarboxylic dianhydride, 98%

CAS: 128-69-8 | C24H8O6 | 392.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 130080250
Price (USD)
50.65
Special offer
Online exclusive
Ends: 28-Dec-2026
59.40
Save 8.75
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
50.65
Special offer
Online exclusive
Ends: 28-Dec-2026
59.40
Save 8.75
Each
Chemical Identifiers
CAS128-69-8
IUPAC Name7,18-dioxaheptacyclo[14.6.2.2²,⁵.0³,¹².0⁴,⁹.0¹³,²³.0²⁰,²⁴]hexacosa-1(22),2(26),3,5(25),9,11,13,15,20,23-decaene-6,8,17,19-tetrone
Molecular FormulaC24H8O6
InChI KeyCLYVDMAATCIVBF-UHFFFAOYSA-N
SMILESO=C1OC(=O)C2=CC=C3C4=CC=C5C(=O)OC(=O)C6=CC=C(C7=CC=C1C2=C37)C4=C56
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark red
Infrared spectrumConforms
Appearance (Form)Powder
Water=<1 % (K.F.)
Assay>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only