9-Phenanthrenecarboxaldehyde, Thermo Scientific Chemicals
9-Phenanthrenecarboxaldehyde, Thermo Scientific Chemicals
9-Phenanthrenecarboxaldehyde, Thermo Scientific Chemicals
Thermo Scientific Chemicals

9-Phenanthrenecarboxaldehyde, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantityPackaging
1301100505 gGlass bottle
Catalog number 130110050
Price (TWD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4707-71-5
IUPAC Namephenanthrene-9-carbaldehyde
Molecular FormulaC15H10O
InChI KeyQECIGCMPORCORE-UHFFFAOYSA-N
SMILESO=CC1=CC2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Form)Powder
Appearance (Color)Light yellow
Melting point99°C to 103°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only