BioLite™ Cell Culture Treated Dishes
Thermo Scientific™

BioLite™ Cell Culture Treated Dishes

Perform general cell culture procedures more economically with Thermo Scientific™ BioLite™ Cell Culture Treated Dishes that are guaranteed to be sterile, non-pyrogenic, and non-cytotoxic.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)
130183150 mm
13018035 mm
13018160 mm
130182100 mm
Catalog number 130183
Price (USD)
393.65
Online exclusive
437.00
Save 43.35
Case of 120
Diameter (Metric):
150 mm
Price (USD)
393.65
Online exclusive
437.00
Save 43.35
Case of 120
Experience optimal performance with wide selections in size, Thermo Scientific™ BioLite™ cell culture treated dishes suit a multitude of experimental needs while providing cost efficiency for a wide selection of applications in your cell culture routine.
• Cell culture treated
• Sterile per ISO 11137, SAL 10-6
• USP Class VI and non-cytotoxic
• Non-pyrogenic

Specifications
Height (Metric) Interior15.4 mm
No. per Case120
No. per Pack10
ShapeRound
Shelf Life2 years
TypeCell Culture Dish
Volume (Metric) Working35 mL
Capacity (Metric)35 mL (Well)
Culture Area148 sq. cm
Diameter (Metric)150 mm
Height (Metric)20 mm
MaterialPolyethylene
SterilitySterile
Surface TreatmentCell Culture
Unit SizeCase of 120

Frequently asked questions (FAQs)

What is the recommended working volume of Nunc adherent cell culture products?

This depends largely on the cell line and working conditions (considerations include headspace for gas exchange, supplements used, and optimal feeding schedule, among others) but we do offer guidelines for development purposes (0.3 to 0.5 mL per cm2) . Note that these volumes are approximate and may have to be modified for best performance. T25 Flask = 7 mL, T75 Flask = 25 mL, T175 Flask = 55 mL, T225 Flask = 70 mL, 300cm2 Flask = 100 mL, 500 cm2 TripleFlask = 200 mL, 35 mm round culture dish = 3 mL, 60 mm round culture dish = 5 mL, 100 mm round culture dish = 12 mL, 150 mm round culture dish = 35 mL, 4 (rectangular) well Multidish = 5 mL, 8 (rectangular) well Multidish = 3 mL, 4 (round) well Multidish = 1 mL, 6 well Multidish = 3 mL, 12 well Multidish = 2 mL, 24 well Multidish = 1 mL, 48 well Multidish = 0.5 mL.